Product Details

SNP ID
rs148950292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33238182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGGACTGCTTGGAAAAGTGAC[A/G]TCAGTGATTCCTGGCCTGAACAACA
Phenotype
MIM: 607412
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BPIFA1 PubMed Links

Gene Details

Gene
BPIFA1
Gene Name
BPI fold containing family A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243193.1 358 Silent Mutation ACA,ACG T96T NP_001230122.1
NM_016583.3 358 Silent Mutation ACA,ACG T96T NP_057667.1
NM_130852.2 358 Silent Mutation ACA,ACG T96T NP_570913.1

View Full Product Details