Product Details

SNP ID
rs149040516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1310474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGATGGAGTGGTCCATGGTGTG[A/G]TGGAGCAGGACTGGAGGCAACCTGG
Phenotype
MIM: 604942
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP1A-SDCBP2 PubMed Links

Gene Details

Gene
FKBP1A-SDCBP2
Gene Name
FKBP1A-SDCBP2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
SDCBP2
Gene Name
syndecan binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199784.1 1016 Silent Mutation CAC,CAT H282H NP_001186713.1
NM_015685.5 1016 Silent Mutation CAC,CAT H197H NP_056500.2
NM_080489.4 1016 Silent Mutation CAC,CAT H282H NP_536737.3
Gene
SNPH
Gene Name
syntaphilin
There are no transcripts associated with this gene.

View Full Product Details