Product Details
- SNP ID
-
rs149059656
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:1537174 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTCACCTGTGAGTCCCAGCAGCCG[A/G]AGGCAGCAGCAGGGTTGGACGAAAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SIRPD
PubMed Links
Gene Details
- Gene
- SIRPD
- Gene Name
- signal regulatory protein delta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178460.2 |
602 |
Silent Mutation |
CTC,CTT |
L186L |
NP_848555.2 |
View Full Product Details