Product Details
- SNP ID
-
rs149138276
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23440293 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGGGATCGGATGCTGGAGAAACC[C/G]CCTGCTGCTGCTGATTGCCCTGGTC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CSTL1
PubMed Links
Gene Details
- Gene
- CSTL1
- Gene Name
- cystatin like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138283.1 |
272 |
Missense Mutation |
CCC,CGC |
P9R |
NP_612140.1 |
View Full Product Details