Product Details

SNP ID
rs149477193
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35542370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCTGCTACTGTTCCTGTCCGAG[C/T]TGCAGTATTACCTCACCACGGAGGT
Phenotype
MIM: 616971
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ERGIC3 PubMed Links

Gene Details

Gene
ERGIC3
Gene Name
ERGIC and golgi 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015966.2 205 Silent Mutation CTG,TTG L46L NP_057050.1
NM_198398.1 205 Silent Mutation CTG,TTG L46L NP_938408.1

View Full Product Details