Product Details

SNP ID
rs149676789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49247911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGTGGTCATAGAAGCTGATGT[A/G]ATTCTCAACCAGACCCAGGTCCTTC
Phenotype
MIM: 616621
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX27 PubMed Links

Gene Details

Gene
DDX27
Gene Name
DEAD-box helicase 27
There are no transcripts associated with this gene.

Gene
ZNFX1
Gene Name
zinc finger NFX1-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021035.2 4286 Missense Mutation CAC,TAC H1705Y NP_066363.1
XM_006723836.1 4286 Missense Mutation CAC,TAC H1705Y XP_006723899.1
XM_011528924.1 4286 Missense Mutation CAC,TAC H1655Y XP_011527226.1
XM_017027971.1 4286 Missense Mutation CAC,TAC H987Y XP_016883460.1

View Full Product Details