Product Details

SNP ID
rs149680323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5938605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTCTCGTGTGATTCTAAAGTG[C/T]TTGCATTAGATTTGAGGCTGGTGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRMT6 PubMed Links

Gene Details

Gene
TRMT6
Gene Name
tRNA methyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281467.1 1673 Missense Mutation AAC,AGC N305S NP_001268396.1
NM_015939.4 1673 Missense Mutation AAC,AGC N475S NP_057023.2
XM_017027875.1 1673 Missense Mutation AAC,AGC N293S XP_016883364.1

View Full Product Details