Product Details

SNP ID
rs149794908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2794294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGCCTCTGTTTGGGAGTCTTGG[C/T]GAGCACGAAATTGCAGGGGAAGGGG
Phenotype
MIM: 609555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPXM1 PubMed Links

Gene Details

Gene
CPXM1
Gene Name
carboxypeptidase X (M14 family), member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184699.1 1953 Missense Mutation NP_001171628.1
NM_019609.4 1953 Missense Mutation NP_062555.1

View Full Product Details