Product Details

SNP ID
rs149956664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45094874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGTCACGGATCCCTGGTTTTAA[C/T]ACATCTGAGGGTGTGGAGGCTCACT
Phenotype
MIM: 602905
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNS1 PubMed Links

Gene Details

Gene
KCNS1
Gene Name
potassium voltage-gated channel modifier subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322799.1 1974 Missense Mutation TAT,TGT Y526C NP_001309728.1
NM_002251.4 1974 Missense Mutation TAT,TGT Y526C NP_002242.2
XM_017027846.1 1974 Missense Mutation TAT,TGT Y527C XP_016883335.1

View Full Product Details