Product Details

SNP ID
rs150270406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49636392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAGGCGTCGTATGTGATGTTTG[C/G]AAAGTAGTTCAGGTTGTTGAGGCCA
Phenotype
MIM: 604016
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
B4GALT5 PubMed Links

Gene Details

Gene
B4GALT5
Gene Name
beta-1,4-galactosyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004776.3 1281 Missense Mutation CCA,GCA P363A NP_004767.1

View Full Product Details