Product Details

SNP ID
rs150281189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:20369169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAGTACCGCTGTCCCGAGTGCG[C/G]CAAGGTCTTCAGCTGCCCGGCCAAC
Phenotype
MIM: 600010
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFAP61 PubMed Links

Gene Details

Gene
CFAP61
Gene Name
cilia and flagella associated protein 61
There are no transcripts associated with this gene.

Gene
INSM1
Gene Name
INSM transcriptional repressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002196.2 1049 Missense Mutation GCC,GGC A301G NP_002187.1

View Full Product Details