Product Details

SNP ID
rs150530248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31607211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCGTAGGATCTGAGGAGTGGA[C/T]GAGTCTCATTACCCAGCTCCTGAGC
Phenotype
MIM: 600349
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ID1 PubMed Links

Gene Details

Gene
ID1
Gene Name
inhibitor of DNA binding 1, HLH protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002165.3 Intron NP_002156.2
NM_181353.2 Intron NP_851998.1
Gene
MIR3193
Gene Name
microRNA 3193
There are no transcripts associated with this gene.

View Full Product Details