Product Details

SNP ID
rs150583052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58313763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTGGTGCCTAGACCAGGGGTG[A/G]GCAAACTGTGGCCTTTTTGCCCGCC
Phenotype
MIM: 612966
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP4R1L PubMed Links

Gene Details

Gene
PPP4R1L
Gene Name
protein phosphatase 4 regulatory subunit 1 like (pseudogene)
There are no transcripts associated with this gene.

Gene
RAB22A
Gene Name
RAB22A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020673.2 Intron NP_065724.1

View Full Product Details