Product Details

SNP ID
rs150719975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:20369788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCCTCCTGCAGGTGCCCGTGCG[A/C]CCGGCCTGCTAGAGCGCGCCCTCCA
Phenotype
MIM: 600010
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CFAP61 PubMed Links

Gene Details

Gene
CFAP61
Gene Name
cilia and flagella associated protein 61
There are no transcripts associated with this gene.

Gene
INSM1
Gene Name
INSM transcriptional repressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002196.2 1668 Silent Mutation CGA,CGC R507R NP_002187.1

View Full Product Details