Product Details

SNP ID
rs150772123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36652109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCTCCTACAAAAGGGAGTTCA[C/T]AGGATTAACAATCTTTCTAATGGCT
Phenotype
MIM: 605273 MIM: 606577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDRG3 PubMed Links

Gene Details

Gene
NDRG3
Gene Name
NDRG family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022477.3 2627 UTR 3 NP_071922.2
NM_032013.3 2627 UTR 3 NP_114402.1
XM_006723837.3 2627 UTR 3 XP_006723900.1
XM_006723838.3 2627 UTR 3 XP_006723901.1
XM_011528925.2 2627 UTR 3 XP_011527227.1
XM_011528926.2 2627 UTR 3 XP_011527228.1
XM_011528927.2 2627 UTR 3 XP_011527229.1
XM_011528928.2 2627 UTR 3 XP_011527230.1
XM_017027978.1 2627 UTR 3 XP_016883467.1
XM_017027979.1 2627 UTR 3 XP_016883468.1
XM_017027980.1 2627 UTR 3 XP_016883469.1
Gene
SLA2
Gene Name
Src like adaptor 2
There are no transcripts associated with this gene.

View Full Product Details