Product Details

SNP ID
rs150946789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45857864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTTCAAGACCTATGAGGACTT[A/C]AAGGAGTGCTTCAGCGCCTACAAAA
Phenotype
MIM: 608123
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACOT8 PubMed Links

Gene Details

Gene
ACOT8
Gene Name
acyl-CoA thioesterase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005469.3 284 Intron NP_005460.2
XM_005260239.3 284 Intron XP_005260296.1
XM_011528480.2 284 Intron XP_011526782.1
Gene
ZSWIM3
Gene Name
zinc finger SWIM-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080752.3 284 Missense Mutation TTA,TTC L13F NP_542790.2

View Full Product Details