Product Details

SNP ID
rs150988771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49248037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCTCCAGCAGGGCCTTAAGCC[A/G]TTCCTGGCTGGTTGCTATTTCCCCT
Phenotype
MIM: 616621
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX27 PubMed Links

Gene Details

Gene
DDX27
Gene Name
DEAD-box helicase 27
There are no transcripts associated with this gene.

Gene
ZNFX1
Gene Name
zinc finger NFX1-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021035.2 4160 Missense Mutation CGG,TGG R1663W NP_066363.1
XM_006723836.1 4160 Missense Mutation CGG,TGG R1663W XP_006723899.1
XM_011528924.1 4160 Missense Mutation CGG,TGG R1613W XP_011527226.1
XM_017027971.1 4160 Missense Mutation CGG,TGG R945W XP_016883460.1

View Full Product Details