Product Details

SNP ID
rs138450273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33549917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATCTCAGAGCAGTCAGAGCAGT[A/C]TGTGGCAGTAATGCCAGAACCATCC
Phenotype
MIM: 138440 MIM: 182465
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GART PubMed Links

Gene Details

Gene
GART
Gene Name
phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase
There are no transcripts associated with this gene.

Gene
MIR6501
Gene Name
microRNA 6501
There are no transcripts associated with this gene.

Gene
SON
Gene Name
SON DNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291411.1 741 Missense Mutation TAT,TCT Y229S NP_001278340.1
NM_001291412.1 741 Intron NP_001278341.1
NM_032195.2 741 Missense Mutation TAT,TCT Y229S NP_115571.2
NM_138927.2 741 Missense Mutation TAT,TCT Y229S NP_620305.2

View Full Product Details