Product Details

SNP ID
rs138494765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30613844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCATAGCCATAGCCCAGGCCTC[C/T]ATAGCCACAGAACCCATAGCCAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP20-1 PubMed Links

Gene Details

Gene
KRTAP20-1
Gene Name
keratin associated protein 20-1
There are no transcripts associated with this gene.

Gene
KRTAP20-4
Gene Name
keratin associated protein 20-4
There are no transcripts associated with this gene.

Gene
KRTAP6-1
Gene Name
keratin associated protein 6-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181602.1 61 Missense Mutation AGA,GGA R21G NP_853633.1

View Full Product Details