Product Details

SNP ID
rs138819156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44498973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCACAGGAGAGCCAGCAGAGGAT[A/G]CTGTTACCAGGGAGCTAGGACAGCC
Phenotype
MIM: 612920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100505727 PubMed Links

Gene Details

Gene
LOC100505727
Gene Name
uncharacterized LOC100505727
There are no transcripts associated with this gene.

Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 2948 UTR 3 NP_001258966.1
NM_144991.2 2948 UTR 3 NP_659428.2
Gene
TSPEAR-AS1
Gene Name
TSPEAR antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details