Product Details

SNP ID
rs138872294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:28539528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGATGAGGTCATACTTGAGTGA[C/G]GTGGGTTTCTAATCCAATGTGACTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00161 PubMed Links

Gene Details

Gene
LINC00161
Gene Name
long intergenic non-protein coding RNA 161
There are no transcripts associated with this gene.

View Full Product Details