Product Details
- SNP ID
-
rs139054863
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:39599489 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGAGCTGGCATAGGGTAGCCCCA[A/G]TGCTGAAGATTCACCAGTGATAAAC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
B3GALT5-AS1
PubMed Links
Gene Details
- Gene
- B3GALT5-AS1
- Gene Name
- B3GALT5 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details