Product Details

SNP ID
rs139497735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30430475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGCTGCCAGAAATCCTGCTACC[A/G]CCCCAGGACCTCCATCCTCTGCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP13-3 PubMed Links

Gene Details

Gene
KRTAP13-3
Gene Name
keratin associated protein 13-3
There are no transcripts associated with this gene.

Gene
KRTAP13-4
Gene Name
keratin associated protein 13-4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181600.1 200 Missense Mutation CAC,CGC H67R NP_853631.1
Gene
KRTAP15-1
Gene Name
keratin associated protein 15-1
There are no transcripts associated with this gene.

View Full Product Details