Product Details

SNP ID
rs140253885
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37065617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTAAAGTTCTTTGGCTGCAAGAA[A/C]GAACATTTGGTTTACTTCACTAATA
Phenotype
MIM: 605938 MIM: 602259
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
LOC105372795 PubMed Links
Additional Information
For this assay, SNP(s) [rs16994704] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC105372795
Gene Name
uncharacterized LOC105372795
There are no transcripts associated with this gene.

Gene
PIGP
Gene Name
phosphatidylinositol glycan anchor biosynthesis class P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320480.1 551 Missense Mutation GTT,TTT V124F NP_001307409.1
NM_016430.3 551 Missense Mutation GTT,TTT V98F NP_057514.2
NM_153681.2 551 Missense Mutation GTT,TTT V148F NP_710148.1
NM_153682.2 551 Missense Mutation GTT,TTT V124F NP_710149.1
XM_005260990.4 551 Intron XP_005261047.1
XM_011529596.2 551 Missense Mutation GTT,TTT V124F XP_011527898.1
XM_017028364.1 551 Missense Mutation GTT,TTT V124F XP_016883853.1
XM_017028365.1 551 Missense Mutation GTT,TTT V98F XP_016883854.1
Gene
TTC3
Gene Name
tetratricopeptide repeat domain 3
There are no transcripts associated with this gene.

View Full Product Details