Product Details

SNP ID
rs140294541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30371620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTAAGAGGGGTTAGCTCACATTG[C/T]TGGAAATGCCTATGATAACAGCTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP13-2 PubMed Links

Gene Details

Gene
KRTAP13-2
Gene Name
keratin associated protein 13-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181621.3 620 UTR 3 NP_853652.1
Gene
MIR4327
Gene Name
microRNA 4327
There are no transcripts associated with this gene.

View Full Product Details