Product Details
- SNP ID
-
rs141195315
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:44330206 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGTTCAGGTCCTGCGGTCACTCGG[C/T]GTGCTCCTGCACCTCTTCCCCACGC
- Phenotype
-
MIM: 603191
MIM: 171860
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C21orf2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11552066] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C21orf2
- Gene Name
- chromosome 21 open reading frame 2
- Gene
- PFKL
- Gene Name
- phosphofructokinase, liver type
There are no transcripts associated with this gene.
View Full Product Details