Product Details

SNP ID
rs141237312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33578422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTTAGGGTGCAAACCACAGTTA[A/G]TAAGCTCCTTATGAACAACCTCCTG
Phenotype
MIM: 611428 MIM: 182465
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DONSON PubMed Links

Gene Details

Gene
DONSON
Gene Name
downstream neighbor of SON
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017613.3 1653 Missense Mutation ACT,ATT T529I NP_060083.1
Gene
SON
Gene Name
SON DNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291411.1 1653 Intron NP_001278340.1
NM_001291412.1 1653 Intron NP_001278341.1
NM_032195.2 1653 Intron NP_115571.2
NM_138927.2 1653 Intron NP_620305.2

View Full Product Details