Product Details

SNP ID
rs142147199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42313575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTTGTTGCACTCCTTGGGGGTGA[C/T]ATGGGGGTAGCCGCAGTCCACCCTG
Phenotype
MIM: 603076 MIM: 600633
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCG1 PubMed Links

Gene Details

Gene
ABCG1
Gene Name
ATP binding cassette subfamily G member 1
There are no transcripts associated with this gene.

Gene
TFF3
Gene Name
trefoil factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003226.3 361 Missense Mutation ATC,GTC I61V NP_003217.3

View Full Product Details