Product Details

SNP ID
rs142778878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:36070343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCATCCGCGCCCTGCGCGACTT[A/C]CTGCGCAAGGAGTACGGGGGCCTGG
Phenotype
MIM: 114830
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CBR1 PubMed Links

Gene Details

Gene
CBR1
Gene Name
carbonyl reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286789.1 420 Silent Mutation TTA,TTC L76F NP_001273718.1
NM_001757.3 420 Silent Mutation TTA,TTC L76F NP_001748.1
Gene
LOC100133286
Gene Name
uncharacterized LOC100133286
There are no transcripts associated with this gene.

Gene
SETD4
Gene Name
SET domain containing 4
There are no transcripts associated with this gene.

View Full Product Details