Product Details
- SNP ID
-
rs143003008
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:32578766 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCACTCTGATTCTTGTGGCCAGC[A/G]TATTTGGGTATTGTCTCTTCATCAG
- Phenotype
-
MIM: 608365
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TCP10L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9622] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TCP10L
- Gene Name
- t-complex 10-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144659.5 |
542 |
Silent Mutation |
TAC,TAT |
Y142Y |
NP_653260.1 |
View Full Product Details