Product Details

SNP ID
rs143382738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32737271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTACCTTGCATTTCTTTTTTC[C/T]ACATCAGAACACCCGATACTATTTC
Phenotype
MIM: 604297
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAXBP1 PubMed Links

Gene Details

Gene
PAXBP1
Gene Name
PAX3 and PAX7 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013329.3 2809 Intron NP_037461.2
NM_016631.3 2809 Silent Mutation GTA,GTG V873V NP_057715.2
XM_006724066.1 2809 Silent Mutation GTA,GTG V838V XP_006724129.1
XM_011529804.2 2809 Intron XP_011528106.1
Gene
PAXBP1-AS1
Gene Name
PAXBP1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SYNJ1
Gene Name
synaptojanin 1
There are no transcripts associated with this gene.

View Full Product Details