Product Details
- SNP ID
-
rs144121064
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:42347628 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAGACCTCCATGACGCACTGATC[C/T]GACTCTGCCATGGGACAGGCACAGC
- Phenotype
-
MIM: 182590
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TFF2
PubMed Links
Gene Details
- Gene
- TFF2
- Gene Name
- trefoil factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005423.4 |
376 |
Silent Mutation |
TCA,TCG |
S78S |
NP_005414.1 |
View Full Product Details