Product Details

SNP ID
rs144227902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41990227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTTCCCAGGTCTTCCTTAGAGT[G/T]TAACTATCATTTATTATATTGACTG
Phenotype
MIM: 616485
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZBTB21 PubMed Links

Gene Details

Gene
ZBTB21
Gene Name
zinc finger and BTB domain containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098402.1 4328 UTR 3 NP_001091872.1
NM_001098403.1 4328 UTR 3 NP_001091873.1
NM_001320729.1 4328 UTR 3 NP_001307658.1
NM_001320731.1 4328 UTR 3 NP_001307660.1
NM_020727.4 4328 UTR 3 NP_065778.3
XM_005261121.3 4328 UTR 3 XP_005261178.1
XM_011529588.2 4328 UTR 3 XP_011527890.1
XM_011529590.2 4328 UTR 3 XP_011527892.1
XM_017028360.1 4328 UTR 3 XP_016883849.1
XM_017028361.1 4328 UTR 3 XP_016883850.1

View Full Product Details