Product Details

SNP ID
rs145241759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:39187200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTCGATGGGGCAGTTTTCTGCT[A/G]CTCTTCCTAATCCAGACATTTTCTG
Phenotype
MIM: 605296
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRWD1 PubMed Links

Gene Details

Gene
BRWD1
Gene Name
bromodomain and WD repeat domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007246.2 7084 Intron NP_001007247.1
NM_018963.4 7084 Silent Mutation AGC,AGT S2263S NP_061836.2
NM_033656.3 7084 UTR 3 NP_387505.1
XM_011529611.1 7084 Silent Mutation AGC,AGT S2215S XP_011527913.1
XM_011529612.2 7084 Silent Mutation AGC,AGT S1983S XP_011527914.1
XM_011529613.1 7084 Silent Mutation AGC,AGT S1274S XP_011527915.1
XM_017028373.1 7084 Silent Mutation AGC,AGT S2176S XP_016883862.1
XM_017028374.1 7084 Silent Mutation AGC,AGT S1274S XP_016883863.1
XM_017028375.1 7084 Intron XP_016883864.1
Gene
PSMG1
Gene Name
proteasome assembly chaperone 1
There are no transcripts associated with this gene.

View Full Product Details