Product Details

SNP ID
rs145561377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29006560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTTCTTCAAAAATGGAAAATT[A/T]CCTTTGTCTTTCCGTTTCATCATTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RWDD2B PubMed Links

Gene Details

Gene
RWDD2B
Gene Name
RWD domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320724.1 1009 Nonsense Mutation AAA,TAA K244* NP_001307653.1
NM_016940.2 1009 Nonsense Mutation AAA,TAA K273* NP_058636.1

View Full Product Details