Product Details

SNP ID
rs145653137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30491780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTAGCAGCCATATCCGTAGCCTC[C/T]GAAGCCAGAGCCATATCCATAGCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP19-2 PubMed Links

Gene Details

Gene
KRTAP19-2
Gene Name
keratin associated protein 19-2
There are no transcripts associated with this gene.

Gene
KRTAP19-3
Gene Name
keratin associated protein 19-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181609.3 178 Missense Mutation NP_853640.1
Gene
KRTAP19-4
Gene Name
keratin associated protein 19-4
There are no transcripts associated with this gene.

View Full Product Details