Product Details

SNP ID
rs146860442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:36461225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACAGGGCCTGGCCAATCTCAAA[C/T]TTCATGCCGCTGGGCAGCAGCGGGT
Phenotype
MIM: 605608
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN14 PubMed Links

Gene Details

Gene
CLDN14
Gene Name
claudin 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146077.1 945 Silent Mutation AAA,AAG K157K NP_001139549.1
NM_001146078.2 945 Silent Mutation AAA,AAG K157K NP_001139550.1
NM_001146079.1 945 Silent Mutation AAA,AAG K157K NP_001139551.1
NM_012130.3 945 Silent Mutation AAA,AAG K157K NP_036262.1
NM_144492.2 945 Silent Mutation AAA,AAG K157K NP_652763.1

View Full Product Details