Product Details

SNP ID
rs147358478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33488892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGTAGTTTCTGCTTTTGATATT[A/C]CATCACTTGTTCAGCAGCACGGTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC28 PubMed Links

Gene Details

Gene
DNAJC28
Gene Name
DnaJ heat shock protein family (Hsp40) member C28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040192.2 951 Missense Mutation NP_001035282.1
NM_001320746.1 951 Nonsense Mutation NP_001307675.1
NM_017833.4 951 Missense Mutation NP_060303.2
Gene
TMEM50B
Gene Name
transmembrane protein 50B
There are no transcripts associated with this gene.

View Full Product Details