Product Details

SNP ID
rs148466782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33276637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAAATGCATGAATACTACCTTGA[C/T]GGAATGTGATTTCTCAAGTCTTTCC
Phenotype
MIM: 123889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL10RB PubMed Links

Gene Details

Gene
IL10RB
Gene Name
interleukin 10 receptor subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000628.4 321 Missense Mutation ACG,ATG T72M NP_000619.3

View Full Product Details