Product Details

SNP ID
rs148929225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41798446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTTCCACGCCGGACCCTCCAG[C/T]GCAGTCCCATGCCGGACACAGCGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRDM15 PubMed Links

Gene Details

Gene
PRDM15
Gene Name
PR domain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040424.2 6502 UTR 3 NP_001035514.1
NM_001282934.1 6502 UTR 3 NP_001269863.1
NM_022115.4 6502 Intron NP_071398.3
XM_006724039.2 6502 UTR 3 XP_006724102.1
XM_006724040.2 6502 UTR 3 XP_006724103.1
XM_011529674.1 6502 UTR 3 XP_011527976.1
XM_011529675.1 6502 UTR 3 XP_011527977.1
XM_011529676.2 6502 UTR 3 XP_011527978.1
XM_011529677.2 6502 UTR 3 XP_011527979.1
XM_011529678.2 6502 UTR 3 XP_011527980.1
XM_011529679.2 6502 UTR 3 XP_011527981.1
XM_011529681.2 6502 UTR 3 XP_011527983.1
XM_011529683.1 6502 UTR 3 XP_011527985.1
XM_017028425.1 6502 UTR 3 XP_016883914.1
XM_017028426.1 6502 UTR 3 XP_016883915.1

View Full Product Details