Product Details

SNP ID
rs149557793
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30430292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATCAGCATGTCCTACAACTGCT[G/T]CTCTAGAAACTTCTCCTCCCGCTCC
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
KRTAP13-3 PubMed Links
Additional Information
For this assay, SNP(s) [rs73356728] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KRTAP13-3
Gene Name
keratin associated protein 13-3
There are no transcripts associated with this gene.

Gene
KRTAP13-4
Gene Name
keratin associated protein 13-4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181600.1 17 Missense Mutation TGC,TTC C6F NP_853631.1

View Full Product Details