Product Details

SNP ID
rs137893581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30292597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTCTCCCCTTCATCTGTTTCCG[C/T]TGCTCCTTCTGGGCCTGCTTGGGTG
Phenotype
MIM: 617034 MIM: 610020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GATSL3 PubMed Links

Gene Details

Gene
GATSL3
Gene Name
GATS protein like 3
There are no transcripts associated with this gene.

Gene
TBC1D10A
Gene Name
TBC1 domain family member 10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204240.1 1411 Silent Mutation CAA,CAG Q442Q NP_001191169.1
NM_031937.2 1411 Silent Mutation CAA,CAG Q435Q NP_114143.1

View Full Product Details