Product Details

SNP ID
rs138070396
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37806433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTTTTGTTGGCTTGTTTGCCA[G/T]GAAGGTAGATGTGGGTGGGGAGAAG
Phenotype
MIM: 607422 MIM: 142708
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
GCAT PubMed Links

Gene Details

Gene
GCAT
Gene Name
glycine C-acetyltransferase
There are no transcripts associated with this gene.

Gene
H1F0
Gene Name
H1 histone family member 0
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005318.3 1327 UTR 3 NP_005309.1

View Full Product Details