Product Details
- SNP ID
-
rs138081500
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:38221556 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGGGAGCGGGAGAGGCCGTGGGC[A/G]CCACCACGCCAGGTGGGCCCAGGCT
- Phenotype
-
MIM: 604877
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAFF
PubMed Links
Gene Details
- Gene
- MAFF
- Gene Name
- MAF bZIP transcription factor F
There are no transcripts associated with this gene.
- Gene
- SNORA92
- Gene Name
- small nucleolar RNA, H/ACA box 92
There are no transcripts associated with this gene.
- Gene
- TMEM184B
- Gene Name
- transmembrane protein 184B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001195071.1 |
1346 |
Silent Mutation |
GGC,GGT |
G379G |
NP_001182000.1 |
NM_001195072.1 |
1346 |
Silent Mutation |
GGC,GGT |
G313G |
NP_001182001.1 |
NM_012264.4 |
1346 |
Silent Mutation |
GGC,GGT |
G379G |
NP_036396.2 |
XM_011530112.2 |
1346 |
Silent Mutation |
GGC,GGT |
G419G |
XP_011528414.2 |
XM_011530113.1 |
1346 |
Silent Mutation |
GGC,GGT |
G391G |
XP_011528415.1 |
XM_011530114.1 |
1346 |
Silent Mutation |
GGC,GGT |
G391G |
XP_011528416.1 |
XM_011530115.2 |
1346 |
Silent Mutation |
GGC,GGT |
G367G |
XP_011528417.1 |
XM_011530116.2 |
1346 |
Intron |
|
|
XP_011528418.2 |
XM_011530117.1 |
1346 |
Silent Mutation |
GGC,GGT |
G325G |
XP_011528419.1 |
XM_011530118.1 |
1346 |
Silent Mutation |
GGC,GGT |
G325G |
XP_011528420.1 |
XM_011530119.1 |
1346 |
Silent Mutation |
GGC,GGT |
G275G |
XP_011528421.1 |
XM_017028754.1 |
1346 |
Silent Mutation |
GGC,GGT |
G424G |
XP_016884243.1 |
XM_017028755.1 |
1346 |
Silent Mutation |
GGC,GGT |
G412G |
XP_016884244.1 |
XM_017028756.1 |
1346 |
Silent Mutation |
GGC,GGT |
G386G |
XP_016884245.1 |
XM_017028757.1 |
1346 |
Silent Mutation |
GGC,GGT |
G362G |
XP_016884246.1 |
XM_017028758.1 |
1346 |
Intron |
|
|
XP_016884247.1 |
XM_017028759.1 |
1346 |
Silent Mutation |
GGC,GGT |
G263G |
XP_016884248.1 |
View Full Product Details