Product Details

SNP ID
rs138569023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41473989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAACTGGGAGATTTTAAGCCAGG[G/T]AGTGATGTGATCTGATCATCTGGGC
Phenotype
MIM: 100850
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACO2 PubMed Links

Gene Details

Gene
ACO2
Gene Name
aconitase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098.2 Intron NP_001089.1
XM_017028812.1 Intron XP_016884301.1
Gene
PHF5A
Gene Name
PHD finger protein 5A
There are no transcripts associated with this gene.

View Full Product Details