Product Details

SNP ID
rs138864587
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:28775358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATGCATTTCTAAATTATTTCTA[A/G]TCTTTTGCTATTACAATCTACCCTG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CCDC117 PubMed Links

Gene Details

Gene
CCDC117
Gene Name
coiled-coil domain containing 117
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284263.1 Intron NP_001271192.1
NM_001284264.1 Intron NP_001271193.1
NM_001284265.1 Intron NP_001271194.1
NM_173510.3 Intron NP_775781.1

View Full Product Details