Product Details

SNP ID
rs138882006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23651883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGGAGACGTGGGGCACAGGCA[A/G]GGTAGCAGGTGAGTCTGGCTGCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GUSBP11 PubMed Links

Gene Details

Gene
GUSBP11
Gene Name
glucuronidase, beta pseudogene 11
There are no transcripts associated with this gene.

View Full Product Details