Product Details
- SNP ID
-
rs140052476
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:24225355 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCACCATGGAGGATGGGGAACGCT[C/T]GCCTGGAACTGGGGGCCAGCACCTT
- Phenotype
-
MIM: 137168
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GGT5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7288201] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GGT5
- Gene Name
- gamma-glutamyltransferase 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001099781.2 |
1714 |
Missense Mutation |
AAG,GAG |
K466E |
NP_001093251.1 |
NM_001099782.2 |
1714 |
Missense Mutation |
AAG,GAG |
K433E |
NP_001093252.1 |
NM_001302464.1 |
1714 |
Missense Mutation |
AAG,GAG |
K433E |
NP_001289393.1 |
NM_001302465.1 |
1714 |
Missense Mutation |
AAG,GAG |
K389E |
NP_001289394.1 |
NM_004121.3 |
1714 |
Missense Mutation |
AAG,GAG |
K465E |
NP_004112.2 |
XM_005261557.2 |
1714 |
Missense Mutation |
AAG,GAG |
K466E |
XP_005261614.1 |
XM_005261558.2 |
1714 |
Missense Mutation |
AAG,GAG |
K465E |
XP_005261615.1 |
XM_011530133.1 |
1714 |
Missense Mutation |
AAG,GAG |
K471E |
XP_011528435.1 |
XM_011530134.1 |
1714 |
Missense Mutation |
AAG,GAG |
K471E |
XP_011528436.1 |
XM_011530135.1 |
1714 |
Missense Mutation |
AAG,GAG |
K439E |
XP_011528437.1 |
XM_011530136.1 |
1714 |
Missense Mutation |
AAG,GAG |
K471E |
XP_011528438.1 |
XM_011530137.2 |
1714 |
Missense Mutation |
AAG,GAG |
K471E |
XP_011528439.1 |
XM_017028768.1 |
1714 |
Missense Mutation |
AAG,GAG |
K433E |
XP_016884257.1 |
XM_017028769.1 |
1714 |
Missense Mutation |
AAG,GAG |
K439E |
XP_016884258.1 |
View Full Product Details