Product Details
- SNP ID
-
rs140164059
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:36141550 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTCACTCCCAATGGTTTGTGTGG[C/T]TTCGTAATAATTATTAAGAAGGGAA
- Phenotype
-
MIM: 607253
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
APOL3
PubMed Links
Gene Details
- Gene
- APOL3
- Gene Name
- apolipoprotein L3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014349.2 |
1830 |
Missense Mutation |
ACC,GCC |
T216A |
NP_055164.1 |
NM_030644.1 |
1830 |
Missense Mutation |
ACC,GCC |
T216A |
NP_085147.1 |
NM_145639.1 |
1830 |
Missense Mutation |
ACC,GCC |
T216A |
NP_663614.1 |
NM_145640.2 |
1830 |
Missense Mutation |
ACC,GCC |
T287A |
NP_663615.1 |
NM_145641.2 |
1830 |
Missense Mutation |
ACC,GCC |
T87A |
NP_663616.1 |
NM_145642.2 |
1830 |
Missense Mutation |
ACC,GCC |
T87A |
NP_663617.1 |
XM_006724324.1 |
1830 |
Missense Mutation |
ACC,GCC |
T216A |
XP_006724387.1 |
XM_006724325.2 |
1830 |
Missense Mutation |
ACC,GCC |
T216A |
XP_006724388.1 |
XM_017028946.1 |
1830 |
Missense Mutation |
ACC,GCC |
T217A |
XP_016884435.1 |
XM_017028947.1 |
1830 |
Missense Mutation |
ACC,GCC |
T217A |
XP_016884436.1 |
XM_017028948.1 |
1830 |
Missense Mutation |
ACC,GCC |
T217A |
XP_016884437.1 |
XM_017028949.1 |
1830 |
Missense Mutation |
ACC,GCC |
T217A |
XP_016884438.1 |
XM_017028950.1 |
1830 |
Missense Mutation |
ACC,GCC |
T217A |
XP_016884439.1 |
XM_017028951.1 |
1830 |
Missense Mutation |
ACC,GCC |
T217A |
XP_016884440.1 |
XM_017028952.1 |
1830 |
Missense Mutation |
ACC,GCC |
T87A |
XP_016884441.1 |
View Full Product Details