Product Details

SNP ID
rs140295594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:16592342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAGGATGGCAGTGGCACACCCC[A/G]TGCACACTGTTTCTCCTTTCATGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCT8L2 PubMed Links

Gene Details

Gene
CCT8L2
Gene Name
chaperonin containing TCP1 subunit 8 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014406.4 Intron NP_055221.1
Gene
TPTEP1
Gene Name
transmembrane phosphatase with tensin homology pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details